missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol, FCC, 99.5%, Spectrum™ Chemical
P1456, 57-55-6, C3H8O2
$474.97 - $10621.11
Chemical Identifiers
| CAS | 57-55-6 |
|---|---|
| Molecular Formula | C3H8O2 |
| Molecular Weight (g/mol) | 76.10 |
| InChI Key | DNIAPMSPPWPWGF-UHFFFAOYNA-N |
| IUPAC Name | propane-1,2-diol |
| SMILES | CC(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605904
|
Spectrum Chemical Mfg Cor
P1456500MLPL |
500 mL | Poly Bottle |
Each for $474.97
|
|
||||
|
18605908
|
Spectrum Chemical Mfg Cor
P1456500MLPLCS6 |
6 x 500 mL | Poly Bottle |
Case of 6 Each for $2,448.26
|
|
||||
|
18605905
|
Spectrum Chemical Mfg Cor
P14564LTPL |
4 L | Poly Bottle |
Each for $1,170.03
|
|
||||
|
18605906
|
Spectrum Chemical Mfg Cor
P14564LTPLCS4 |
4 x 4 L | Poly Bottle |
Case of 4 Each for $3,752.74
|
|
||||
|
18605907
|
Spectrum Chemical Mfg Cor
P145620LT |
20 L | Poly Drum |
Each for $4,024.80
|
|
||||
|
18605909
|
Spectrum Chemical Mfg Cor
P1456200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||