missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol, FCC, 99.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Propylene Glycol, FCC is used as a humectant, solvent, and a preservative in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Ignition Residue | 7e-005 |
| Packaging | Poly Bottle |
| Quantity | 500 mL |
| Percent Purity | 99.5% |
| Grade | FCC |