missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, FCC, 99.5%, Spectrum™ Chemical

P1456, 57-55-6, C3H8O2

Supplier:  Spectrum Chemical Mfg Cor P14564LTPL

 View more versions of this product

Catalog No. 18605905


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Propylene Glycol, FCC is used as a humectant, solvent, and a preservative in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

57-55-6
7e-005
C3H8O2
DNIAPMSPPWPWGF-UHFFFAOYNA-N
propane-1,2-diol
99.5%
1
Poly Bottle
4 L
CC(O)CO
76.10
FCC

Chemical Identifiers

57-55-6
76.10
propane-1,2-diol
C3H8O2
DNIAPMSPPWPWGF-UHFFFAOYNA-N
CC(O)CO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions