missing translation for 'onlineSavingsMsg'
Learn More

Propylbenzene 99.0+%, TCI America™

Catalog No. p-7134658
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
P052325ML 25 mL
P0523100ML 100 mL
P0523500ML 500 mL
3 options

Catalog No. P052325ML

Supplier: TCI America P052325ML

Chemical Identifiers

CAS 103-65-1
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009377
InChI Key ODLMAHJVESYWTB-UHFFFAOYSA-N
Synonym n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
PubChem CID 7668
ChEBI CHEBI:42630
IUPAC Name propylbenzene
SMILES CCCC1=CC=CC=C1

Specifications

Melting Point -99°C
Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 2364
Formula Weight 120.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Propylbenzene
EINECSNumber (3)-0021
RTECSNumber DA8750000
TSCA Yes