accessibility menu, dialog, popup

UserName

CH3

CAS RN 103-65-1

IUPAC Name: propylbenzene
Synonyms: propylbenzene N-propylbenzene isocumene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: ODLMAHJVESYWTB-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)