missing translation for 'onlineSavingsMsg'
Learn More

Propylbenzene 99.0+%, TCI America™

Catalog No. P052325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P052325ML 25 mL
P0523100ML 100 mL
P0523500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P052325ML Supplier TCI America Supplier No. P052325ML

Chemical Identifiers

CAS 103-65-1
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009377
InChI Key ODLMAHJVESYWTB-UHFFFAOYSA-N
Synonym n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
PubChem CID 7668
ChEBI CHEBI:42630
IUPAC Name propylbenzene
SMILES CCCC1=CC=CC=C1

Specifications

Melting Point -99°C
Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 2364
Formula Weight 120.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Propylbenzene
EINECSNumber (3)-0021
RTECSNumber DA8750000
TSCA Yes