missing translation for 'onlineSavingsMsg'
Learn More

Propylbenzene, 98%

Catalog No. AC418430250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418430250 25 g Glass bottle
AC418431000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418430250 Supplier Thermo Scientific Chemicals Supplier No. 418430250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-65-1 | C9H12 | 120.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-65-1
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009377
InChI Key ODLMAHJVESYWTB-UHFFFAOYSA-N
Synonym n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
PubChem CID 7668
ChEBI CHEBI:42630
IUPAC Name propylbenzene
SMILES CCCC1=CC=CC=C1

Specifications

Melting Point -99°C
Color Yellow
Density 0.8600g/mL
Boiling Point 158°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.493
Linear Formula C6H5CH2CH2CH3
Quantity 25 g
Beilstein 05, 390
Specific Gravity 0.86
Merck Index 15, 7957
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: miscible with ethanol,ether and acetone
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Propylbenzene
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement:
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-132-9

RUO – Research Use Only