missing translation for 'onlineSavingsMsg'
Learn More

Propyl Salicylate 98.0+%, TCI America™

Catalog No. S044125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S044125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S044125G Supplier TCI America Supplier No. S044125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 607-90-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00045763
InChI Key LZFIOSVZIQOVFW-UHFFFAOYSA-N
Synonym 2-Hydroxybenzoic Acid Propyl Ester, Salicylic Acid Propyl Ester, Propyl 2-Hydroxybenzoate
PubChem CID 69092
IUPAC Name propyl 2-hydroxybenzoate
SMILES CCCOC(=O)C1=CC=CC=C1O

Specifications

Color Colorless
Boiling Point 246°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Salicylate
EINECSNumber (3)-1585
TSCA Yes