accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 607-90-9

IUPAC Name: propyl 2-hydroxybenzoate
Synonyms: propyl 2-hydroxybenzoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: LZFIOSVZIQOVFW-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results