missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl 4-Hydroxybenzoate 99.0+%, TCI America™
Supplier: TCI America P195525G
Specifications
| Propyl 4-Hydroxybenzoate [for Biochemical Research] | |
| 98°C | |
| C10H12O3 | |
| 25 g | |
| QELSKZZBTMNZEB-UHFFFAOYSA-N | |
| propyl 4-hydroxybenzoate | |
| 7175 | |
| 180.20 | |
| Crystalline Powder |
| 94-13-3 | |
| White | |
| MFCD00002354 | |
| propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept | |
| CCCOC(=O)C1=CC=C(C=C1)O | |
| 180.203 | |
| CHEBI:32063 | |
| ≥99.0% (T) |
Chemical Identifiers
| 94-13-3 | |
| 180.203 | |
| QELSKZZBTMNZEB-UHFFFAOYSA-N | |
| 7175 | |
| propyl 4-hydroxybenzoate |
| C10H12O3 | |
| MFCD00002354 | |
| propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept | |
| CHEBI:32063 | |
| CCCOC(=O)C1=CC=C(C=C1)O |
Safety and Handling
EINECSNumber : (3)-1585
RTECSNumber : DH2800000
TSCA : Yes