missing translation for 'onlineSavingsMsg'
Learn More

Propiolic Acid 97.0+%, TCI America™

Supplier:  TCI America P049725G

 View more versions of this product

Catalog No. P049725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Propiolic Acid
9°C
58°C
MFCD00004360
2920
UORVCLMRJXCDCP-UHFFFAOYSA-N
prop-2-ynoic acid
10110
70.05
Liquid
471-25-0
Yellow
C3H2O2
25 g
propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
OC(=O)C#C
70.05
CHEBI:33199
≥97.0% (GC,T)

Chemical Identifiers

471-25-0
70.05
UORVCLMRJXCDCP-UHFFFAOYSA-N
10110
prop-2-ynoic acid
C3H2O2
MFCD00004360
propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
CHEBI:33199
OC(=O)C#C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1086

RTECSNumber : UD9300000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions