missing translation for 'onlineSavingsMsg'
Learn More

Propiolic Acid 97.0+%, TCI America™

$132.35 - $579.71

Chemical Identifiers

CAS 471-25-0
Molecular Formula C3H2O2
Molecular Weight (g/mol) 70.05
MDL Number MFCD00004360
InChI Key UORVCLMRJXCDCP-UHFFFAOYSA-N
Synonym propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
PubChem CID 10110
ChEBI CHEBI:33199
IUPAC Name prop-2-ynoic acid
SMILES OC(=O)C#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P04975G
View Documents
TCI America
P04975G
5 g
Each for $132.35
Only null left
 
P049725G
View Documents
TCI America
P049725G
25 g
Each for $579.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

471-25-0
70.05
UORVCLMRJXCDCP-UHFFFAOYSA-N
10110
prop-2-ynoic acid
C3H2O2
MFCD00004360
propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
CHEBI:33199
OC(=O)C#C

Specifications

471-25-0
Yellow
C3H2O2
5 g
propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
OC(=O)C#C
70.05
CHEBI:33199
≥97.0% (GC,T)
Propiolic Acid
9°C
58°C
MFCD00004360
2920
UORVCLMRJXCDCP-UHFFFAOYSA-N
prop-2-ynoic acid
10110
70.05
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1086

RTECSNumber : UD9300000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'