missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ PA1 HPLC Columns

Catalog No. DX039657
Change view
Click to view available options
Length (Metric):
50 mm
250 mm
Diameter (Metric):
4 mm
9 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
DX039657 50 mm 4 mm
DX039658 250 mm 4 mm
DX040137 250 mm 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. DX039657 Supplier Thermo Scientific™ Supplier No. 039657

Efficiently analyze and purify hydrophilic anionic proteins and peptides with ProPac™ PA1 LC Columns.

Efficiently analyze and purify hydrophilic anionic proteins and peptides with Thermo Scientific™ ProPac™ PA1 LC Columns. These strong anion-exchange columns provide high-resolution separations of proteins with pI values from 3 to 11.

The columns can also resolve some proteins and peptides that are highly cationic and have very high isoelectric points.

  • Good for hydrophilic proteins and peptides
  • Ideal for high-resolution separations of proteins with pI values from 3 to 11
  • Pellicular packing ensures high-efficiency and fast mass transport

Specifications

Column Format Guard Column
Diameter (Metric) 4 mm
Flow Rate 0.9 to 1.5 mL/min
For Use With BioLC Chromatography System
Length (Metric) 50 mm
Max. Pressure 5000 psi (345 bar)
Particle Size 10 μm
Pore Size Nonporous
Quantity Each
Stationary Phase Alkyl Quaternary Amine
Product Line ProPac
Packing Material Spherical, Nonporous Polymer
Column Type Ion Exchange
Phase Ion Exchange
Material PEEK
Green Features FSC
Product Type Guard Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.