Learn More
Thermo Scientific™ ProPac™ HIC-10 HPLC Columns
Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns.
Supplier: Thermo Scientific™ 063665
Description
Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns. Developed with multi-functional ligand attachments, these silica-based columns provide exceptional hydrolytic stability under the highly aqueous conditions used in HIC.
High-efficiency Columns for the Separation of Large Biomolecules
- High protein loading capacity for protein purification applications
- Wide range of analytical and preparative applications, including the separation of bovine serum proteins, snake venom proteins, and monoclonal antibodies (MAbs)
- Based on 5μm ultra-high purity spherical silica gel particles with 300Å pores
Hydrophobic interaction chromatography separates biomolecules in a decreasing salt gradient, based on differences in surface hydrophobicity. This method preserves the biological activity of proteins. The HIC separation mechanism is complementary to those of ion-exchange and gel filtration chromatography.
Specifications
| ± | |
| 7.8 mm | |
| 3500 psi (240 bar) | |
| 2.5 to 7.5 | |
| Each | |
| 40°C | |
| ProPac | |
| Hydrophobic Interaction | |
| Stainless Steel | |
| Bio-LC |
| Analytical Column | |
| 75 mm | |
| 5 μm | |
| 300 Å | |
| 100 m²/g | |
| Amide/Ethyl | |
| Spherical, Fully Porous, Ultrapure Silica | |
| Hydrophobic Interaction | |
| Analytical Column | |
| Responsibly packaged |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.