missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ HIC-10 HPLC Columns

Catalog No. dx063653
Change view
missing translation for 'orderingAttributeHoverText'
Length (Metric):
100 mm
250 mm
75 mm
Diameter (Metric):
2.1 mm
4.6 mm
7.8 mm
Catalog No. Length (Metric) Diameter (Metric)
DX063653 100 mm 2.1 mm
DX063655 100 mm 4.6 mm
DX063665 75 mm 7.8 mm
DX074197 250 mm 4.6 mm
4 options

Catalog No. DX063653

Supplier: Thermo Scientific™ 063653

Only null left

Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns.

Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns. Developed with multi-functional ligand attachments, these silica-based columns provide exceptional hydrolytic stability under the highly aqueous conditions used in HIC.

High-efficiency Columns for the Separation of Large Biomolecules

  • High protein loading capacity for protein purification applications
  • Wide range of analytical and preparative applications, including the separation of bovine serum proteins, snake venom proteins, and monoclonal antibodies (MAbs)
  • Based on 5μm ultra-high purity spherical silica gel particles with 300Å pores

Hydrophobic interaction chromatography separates biomolecules in a decreasing salt gradient, based on differences in surface hydrophobicity. This method preserves the biological activity of proteins. The HIC separation mechanism is complementary to those of ion-exchange and gel filtration chromatography.

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 100 mm
Max. Pressure 3500 psi (240 bar)
Particle Size 5 μm
pH 2.5 to 7.5
Pore Size 300 Å
Quantity Each
Surface Area 100 m²/g
Temperature 40°C
Stationary Phase Amide/Ethyl
Product Line ProPac
Packing Material Spherical, Fully Porous, Ultrapure Silica
Column Type Hydrophobic Interaction
Phase Hydrophobic Interaction
Material Stainless Steel
Type Analytical Column
Technique Bio-LC
Green Features Responsibly packaged
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.