missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ Elite WCX HPLC Column

Catalog No. p-8557023
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
250 mm
Diameter (Metric):
2 mm
4 mm
Quantity:
1 Column
3 Columns/1 Lot
3 Columns/3 Lots
Catalog No. Length (Metric) Diameter (Metric) Quantity
03251448 50 mm 2 mm 1 Column
03251449 150 mm 2 mm 1 Column
03251450 250 mm 2 mm 1 Column
03251452 150 mm 4 mm 1 Column
03251453 250 mm 4 mm 1 Column
03251454 150 mm 4 mm 3 Columns/1 Lot
03251457 150 mm 4 mm 3 Columns/3 Lots
7 options

Catalog No. 03251448

Supplier: Thermo Scientific™ 303028

Only null left

This column provides consistent separations of therapeutic proteins including monoclonal antibodies, antibody drug conjugates, biosimilars and charge variants.

Discover the unique Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column. Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants.

The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.

Product Highlights
  • Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
  • High efficiency with reproducible separa-tions
  • High recovery with low carry-over
  • Wide pH operating range: 2 – 12
  • High temperature stability: up to 60°C
  • High throughput
  • Compatible with CX-1 pH Gradient Buffers

Specifications

Column Format Analytical Column
Diameter (Metric) 2 mm
Flow Rate 0.1 to 0.8 mL/min
Length (Metric) 50 mm
Max. Pressure 4500 psi (310 bar)
Mobile Phase Compatibility Recommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O
Particle Size 5 μm
pH 2 to 12
Pore Size Nonporous
Quantity 1 Column
Temperature 10°C to 60°C
Stationary Phase Carboxylate
Product Line ProPac
Packing Material Nonporous DVB Particles with Hydrophilic Coating
Column Type Ion Exchange
Phase Ion Exchange
Technique Bio-LC
Green Features Responsibly packaged
Show More Show Less