missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Wizard™ SV Genomic DNA Purification System
The Wizard™ SV Genomic DNA Purification System provides a fast, membrane-based method for preparing genomic DNA from cultured cells and tissue, including mouse tails.
$237.91 - $3,158.04
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| For Use With (Application) | Permits fast, simple preparation of genomic DNA without centrifugation of the lysate prior to purification |
Includes: Genomic DNA purification kit: Collection tubes, 0.5M EDTA (pH 8.0), Nuclease-free water, Nuclei lysis solution, RNase A solution, Wizard SV lysis buffer, Wizard SV minicolumns, Wizard SV wash solution
Genomic DNA Purification start-up kit (110V): Wizard SV Genomic DNA Purification System, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold (20 sample capacity), vacuum adapters
| Catalog Number | Mfr. No. | No. of Reactions | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Product Type | Price | Quantity & Availability | ||||
PRA2360
|
Promega
A2360 |
50 Preps | Purification System |
Each for $237.91
|
|
||||
PRA6770
|
Promega
A6770 |
50 preps | Purification Start-up Kit |
Each for $3,158.04
|
|
||||
PRA2361
|
Promega
A2361 |
250 Preps | Purification System |
Each for $726.07
|
|
||||
Description
- Extracts DNA from Tissues or Cells
- Isolate DNA from cultured cells in 20 minutes
- Choice of spin or vacuum purification protocols
Specifications
| 15°C to 30°C | |
| Permits fast, simple preparation of genomic DNA without centrifugation of the lysate prior to purification |