missing translation for 'onlineSavingsMsg'
Learn More

Promega Wizard™ SV Gel and PCR Cleanup System

Catalog No. PRA9285
Encompass
Change view
Click to view available options
No. of Reactions:
1000 Preps
250 Preps
50 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRA9285 1000 Preps
PRA9282 250 Preps
PRA9281 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRA9285 Supplier Promega Supplier No. A9285
Only null left

Includes

Wizard SV Minicolumns, collection tubes, membrane binding solution, membrane wash solution, nuclease-free water

Requires

Vacuum protocol requires vacuum adapters (A1331, sold separately).

The Wizard™ SV Gel and PCR Clean-Up System is designed to purify DNA fragments of 100bp-10kb from standard or low-melting agarose gels or to purify products directly from PCR or restriction digests.

  • Purifies DNA Fragments from Gels or PCR in 15 Minutes
  • Choice of spin- or vacuum-based protocols
  • Purifies 100bp to 10kb DNA fragments

Specifications

Content And Storage 15°C to 30°C
Format Spin or Vacuum
For Use With (Application) Purifies DNA from agarose gels or PCR reactions in as little as 15 min.
Includes 4 x 100mL Membrane binding solution, 4 x 75mL membrane wash solution, 4 x 13mL Nuclease-free Water, 4 x 250 Wizard™ SV minicolumns, 4 x 250 2mL collection tubes
No. of Reactions 1000 Preps
Product Type System