missing translation for 'onlineSavingsMsg'
Learn More

Promega Wizard™ SV 96 Plasmid DNA Purification System

Catalog No. PRA2258
Encompass
Change view
Click to view available options
No. of Reactions:
1 x 96 Preps
100 x 96 preps
5 x 96 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRA2258 100 x 96 preps
PRA2250 1 x 96 Preps
PRA2255 5 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRA2258 Supplier Promega Supplier No. A2258

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

96-well deep-well plate (except A2258); lysate-clearing plate; DNA-binding plate; DNA elution plate (except A2258); plate sealers (except A2258); cell resuspension solution; cell lysis solution; neutralization solution; wash solution; alkaline protease solution (except A2258); nuclease-free water (except A2258)

Requires

Systems require a vacuum pump capable of generating 15-20 in. of Hg or equivalent.

The Wizard™ SV 96 and SV 9600 Plasmid DNA Purification Systems provide a simple and reliable method for the rapid isolation of plasmid DNA using a silica-membrane, 96-well, high-throughput format.

  • Silica Membrane-Based, 96-Well, High-Throughput Plasmid Purification
  • 60-minute protocol
  • Manual and automated formats
  • Spin or vacuum-based methods

U.S. Patent 5,981,235, Australian Patent 729932.

Specifications

Content And Storage 15°C to 30°C
Includes 3.2L Wizard™ SV 96 Cell Resuspension Solution, 3.2L Wizard™ SV 96 Cell Lysis Solution, 8.5L Wizard™ SV 96 Neutralization Solution, 7.4L Column Wash Solution (CWA), 100 Lysate Clearing Plates, 100 Binding Plates
No. of Reactions 100 x 96 preps