Learn More
Promega Wizard™ Plus SV Minipreps DNA Purification System
A silica membrane-based system for simple, rapid isolation of plasmid DNA from 1-10ml E. coli cultures. The miniprep procedure can be completed in 45 minutes or less.
Supplier:  Promega A6760
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. PRA6760
Il en reste null
Comprend:
  • DNA purification kit: Cell resuspension solution, cell lysis solution, neutralization solution, column wash solution, alkaline protease solution, nuclease-free water, miniprep spin columns, collection tubes with or without vacuum adapters
  • DNA purification start-up kit (110V): Wizard™ Plus SV Minipreps DNA Purification System, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold, vacuum adapters
Description
Description
  • 1-20μg High-Quality Plasmid DNA from 1-10mL Cultures
  • Process up to 20 minipreps in 45 minutes
  • Choice of spin or vacuum purification formats
  • No phenol extractions or precipitations required
Spécifications
Identifiants chimiques
15°C to 30°C
Spin or Vacuum
Produces high yield plasmid DNA of consistent quality from bacterial cultures
1 to 20μg
Spécifications
15°C to 30°C
DNA purification system
Wizard Plus SV Minipreps DNA purification system (3 × 250 preps), Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold, 20 Vacuum adapters
Start-Up Kit, 110V
110V
Spin or Vacuum
Produces high yield plasmid DNA of consistent quality from bacterial cultures
3 x 250 preps
1/Ea.
1 to 20μg
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
U.S. Patent 5,981,235, Australian Patent 729932.