missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Wizard™ Plus DNA Purification Systems
A simple, reliable, silica-resin-based method for rapid isolation of plasmid DNA on a miniprep, midiprep, maxiprep, or megaprep scale.
$243.61 - $733.68
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| For Use With (Application) | Purifies plasmid DNA from culture |
Includes: Appropriate Wizard columns and DNA purification resin, cell resuspension solution, cell lysis solution, neutralization solution, column wash solution
| Catalog Number | Mfr. No. | No. of Reactions | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Product Type | Price | Quantity & Availability | ||||
PRA7100
|
Promega
A7100 |
50 Preps | Minipreps System |
Each for $243.61
|
|
||||
PRA7270
|
Promega
A7270 |
10 Preps | Maxipreps System |
Each for $497.69
|
|
||||
PRA7300
|
Promega
A7300 |
5 Preps | Megapreps System |
Each for $438.69
|
|
||||
PRA7640
|
Promega
A7640 |
25 Preps | Midipreps System |
Each for $383.49
|
|
||||
PRA7510
|
Promega
A7510 |
250 Preps | Minipreps System |
Each for $733.68
|
|
||||
PRA7500
|
Promega
A7500 |
100 Preps | Minipreps System |
Each for $328.31
|
|
||||
Description
- Each system is quality tested in fluorescent sequencing reactions to ensure read lengths of at least 500 base pairs with 98% accuracy
- Proprietary silica-based resin rehydrates rapidly, allowing fast, reproducible elution of DNA using water or TE Fast, manual format allows minipreps to be completed in as little as 15 minutes, midipreps in 90 minutes, and maxi- and megapreps in three hours
- Safe, convenient protocol eliminates the need for phenol extraction and ethanol precipitation
- Purified DNA can be used directly in most molecular biology applications
- Minipreps feature disposable minicolumns with Luer-Lok™ tops for attachment of a 3mL syringe (not included); larger preps use columns that require the use of a vacuum manifold (not included)
Specifications
| 15°C to 30°C | |
| Purifies plasmid DNA from culture |