missing translation for 'onlineSavingsMsg'
Learn More

Promega Wizard™ Genomic DNA Purification Kits

An easy, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.

$259.79 - $1,567.28

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Isolate genomic DNA from whole blood, cultured cells, yeast, mouse tissue or numerous other cell sources
Product Type Genomic DNA Purification Kit
Yield 5 to 15μg/300μL
View More Specs

Includes: Cell lysis solution, nuclei lysis solution, protein precipitation solution, DNA rehydration solution, RNase solution‡

Products 3
Catalog Number Mfr. No. No. of Reactions Sample Volume Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Sample Volume Price Quantity & Availability  
PRA1620
Encompass
View Documents Promotion Details
Promega
A1620
100 Preps 10mL
Each for $1,567.28
Only null left
 
PRA1120
Encompass
View Documents Promotion Details
Promega
A1120
100 Preps 300μL
Each for $259.79
Only null left
 
PRA1125
Encompass
View Documents Promotion Details
Promega
A1125
500 Preps 300μL
Each for $633.76
Only null left
 
Description

Description

  • Manual, Solution-Based DNA Extraction Method
  • DNA from blood, animal and plant cells, bacteria or yeast
  • 60-minute, scalable protocol
  • Kit size based on number of isolations from whole blood, other sample types may vary

† To fully rehydrate DNA, allow an additional hour at 65°C (149°F) or overnight at room temperature.

‡ PR-A1620 does not include RNase Solution.

Specifications

Specifications

15°C to 30°C
Isolate genomic DNA from whole blood, cultured cells, yeast, mouse tissue or numerous other cell sources
Genomic DNA Purification Kit
5 to 15μg/300μL
Videos
SDS
Documents

Documents