missing translation for 'onlineSavingsMsg'
Learn More

Promega Western Blue™ Stabilized Substrate for Alkaline Phosphatase

Catalog No. PRS3841
Encompass
Change view
Click to view available options
Quantity:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRS3841 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRS3841 Supplier Promega Supplier No. S3841

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stable, ready-to-use substrate for Western blots and immunoscreening.

  • Ready-to-use formulation that does not require dilution or reagent mixing
  • As sensitive as other commercially available BCIP/NBT formulations and reagents
  • Stable for one year at room temperature

Specifications

For Use With (Application) Western blots and immunoscreening
Quantity 100mL
Substrate Type AP (Alkaline Phosphatase) Substrate
pH 9.8
Storage Requirements 15 to 30°C
Substrate Quantity 100mL