Learn More
Promega VivoGlo™ In Vivo Imaging Substrates DFS Item
VivoGlo™ Luciferin is the potassium salt of D-luciferin, the firefly luciferase substrate capable of generating light when a suitable model is used.
Supplier:  Promega P1043
 View more versions of this product
Catalog No. PRP1043
Only null left
Explore available promotions
Description
Description
  • Firefly Luciferase Substrate for Use in Animal Imaging Studies
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings
Specifications
Specifications
In-vivo bioluminescence imaging applications
VivoGlo Luciferin
1g
1g
−30 to −10°C
Product Suggestions
Product Suggestions