missing translation for 'onlineSavingsMsg'
Learn More

Promega VivoGlo™ In Vivo Imaging Substrates

Catalog No. PRP1042
Encompass
Change view
Click to view available options
Substrate Quantity:
50mg
250mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Substrate Quantity
PRP1042 250mg
PRP1041 50mg
PRP1043 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRP1042 Supplier Promega Supplier No. P1042

VivoGlo™ Luciferin is the potassium salt of D-luciferin, the firefly luciferase substrate capable of generating light when a suitable model is used.

  • Firefly Luciferase Substrate for Use in Animal Imaging Studies
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings

Specifications

For Use With (Application) In-vivo bioluminescence imaging applications
Quantity 250mg
Substrate Type VivoGlo Luciferin
Storage Requirements −30 to −10°C
Substrate Quantity 250mg