missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega ViviRen™ In Vivo Renilla Luciferase Substrate
Description
- Engineered Coelenterazine-Based Substrate for Animal Imaging
- High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
- Available in multiple sizes to accommodate a variety of experimental settings
Specifications
Specifications
| For Use With (Application) | In-vivo bioluminescence imaging applications |
| Quantity | 3.7mg |
| Substrate Type | Renilla Luciferase Substrate |
| Storage Requirements | −30 to −10°C |
| Substrate Quantity | 3.7mg |