missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Viral ToxGlo™ Assay Systems
The Viral ToxGlo™ Assay is a simple, quantifiable method of determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions. The assay measures cellular ATP as a surrogate measure of host cell viability.
$247.71 - $1,382.16
Specifications
| Detection Method | Luminescence |
|---|---|
| For Use With (Application) | Determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions |
| Kit Contents | ATP detection buffer and ATP detection substrate |
| Content And Storage | -30°C to -10°C. |
Alert:
This method is only useful for viruses that produce cytotoxicity and CPE. Not for use in diagnostic procedures.
Description
- Determine Viral CPE or Monitor Compounds for Antiviral Activity
- Easy, simple protocol measures ATP
- Quantifiable assay for measuring viral-induced cytopathic effect (CPE)
- Compatible with high-throughput screening
Specifications
| Luminescence | |
| Determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions | |
| ATP detection buffer and ATP detection substrate | |
| -30°C to -10°C. |
Research use only.