missing translation for 'onlineSavingsMsg'
Learn More

Promega Viral ToxGlo™ Assay Systems

Catalog No. PRG8941
Encompass
Change view
Click to view available options
Quantity:
1 x 10 mL
10 x 10 mL
1 x 100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8941 1 x 10 mL
PRG8942 10 x 10 mL
PRG8943 1 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This method is only useful for viruses that produce cytotoxicity and CPE. Not for use in diagnostic procedures.
Catalog No. PRG8941 Supplier Promega Supplier No. G8941
Only null left
This method is only useful for viruses that produce cytotoxicity and CPE. Not for use in diagnostic procedures.

The Viral ToxGlo™ Assay is a simple, quantifiable method of determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions. The assay measures cellular ATP as a surrogate measure of host cell viability.

  • Determine Viral CPE or Monitor Compounds for Antiviral Activity
  • Easy, simple protocol measures ATP
  • Quantifiable assay for measuring viral-induced cytopathic effect (CPE)
  • Compatible with high-throughput screening

Specifications

Quantity 1 x 10 mL
Content And Storage ATP detection buffer and ATP detection substrate. Store at -30°C to -10°C.
Detection Method Luminescence
For Use With (Application) Determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions

Research use only.