Learn More
Promega Viral ToxGlo™ Assay Systems
The Viral ToxGlo™ Assay is a simple, quantifiable method of determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions. The assay measures cellular ATP as a surrogate measure of host cell viability.
Supplier: Promega G8942
Alert:
This method is only useful for viruses that produce cytotoxicity and CPE. Not for use in diagnostic procedures.
Description
- Determine Viral CPE or Monitor Compounds for Antiviral Activity
- Easy, simple protocol measures ATP
- Quantifiable assay for measuring viral-induced cytopathic effect (CPE)
- Compatible with high-throughput screening
Chemical Identifiers
| Luminescence | |
| Determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions | |
| ATP detection buffer and ATP detection substrate | |
| -30°C to -10°C. |
Specifications
| 10 x 10 mL | |
| Determining viral-induced cytopathic effects (CPE) in host cells caused by lytic virions | |
| -30°C to -10°C. |
| Luminescence | |
| ATP detection buffer and ATP detection substrate |
Research use only.