missing translation for 'onlineSavingsMsg'
Learn More

Promega Vac-Man™ Vacuum Manifold

For use with any Wizard™ DNA Purification System or SV Total RNA Isolation system

$209.35

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Wizard™ Nucleic Acid purification products
For Use With (Application) Simple and reliable method for rapid isolation of plasmid DNA
Quantity 1/Ea.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRA7231
Encompass
View Documents Promotion Details
Promega
A7231
1/Ea.
Each for $209.35
Only null left
 
Description

Description

  • 20 Sample capacity Supplied with 20 individual One-Way Luer-Lok™ stopcocks for individual port control

Specifications

Specifications

15°C to 30°C
Wizard™ Nucleic Acid purification products
Simple and reliable method for rapid isolation of plasmid DNA
1/Ea.
Videos
SDS
Documents

Documents