missing translation for 'onlineSavingsMsg'
Learn More

Promega Trypsin Gold, Mass Spectrometry Grade

Catalog No. PRV5280
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV5280 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV5280 Supplier Promega Supplier No. V5280

missing translation for 'validMainframeMsg'

Trypsin Gold, Mass Spectrometry Grade, provides maximum specificity for mass spectrometry.

  • Maximum Digest Specificity with Extreme Resistance to Autolytic Digestion
  • Each lot qualified by mass spectrometry to ensures compatibility with your applications/instrumentation
  • Chymotrypsin activity eliminated for distinct, consistent data
  • Digest in gel, or in solution

Specifications

Color White
pH 7.7
For Use With (Application) Trypsin digestion of Proteins from 1 or 2-D Gel, tryptic digestion of Proteins in-solution
Storage Buffer 50mM Acetic Acid Resuspension Buffer
Content And Storage -30°C to -10°C
Quantity 100 μg
Product Type Trypsin Gold
Inhibitors Antipain (50μg/mL), Antithrombin (1U/mL), APMSF (0.01 to 0.04mg/mL), Aprotinin (0.06 to 2μg/mL), Leupeptin (0.5μg/mL), PMSF (17 to 170μg/mL), TLCK (37 to 50μg/mL)
Form Lyophilized Powder