missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega TNT™ SP6 High-Yield Protein Expression System
Description
Includes
TNT SP6 High-Yield Master Mix, Nuclease-free water
- Coupled Transcription/Translation for Soluble, Full-Length Protein in Only Two Hours
- Express soluble, full-length protein from plasmid or PCR-generated templates quickly
- Generate 10- to 20-fold more protein (10-100μg/ml) than with other cell-free systems
Specifications
Specifications
| Kit Contents | TnT™ SP6 high-yield master mix, Nuclease-free Water |
| Test Time | 2 hr. |
| Yield | 10 to 100 μg/mL |
| Type | Wheat Germ Protein Expression System |
| Sufficient For | 10 Reactions |
| Quantity | 10 reactions |
| Content And Storage | <-65°C |
- U.S. Patent 5,324,637; Australian Patent 660329 and other patents.
- For Research Use. Any use of the product for diagnostics requiring clearance or approval by the FDA may require a license under Mayo Clinic U.S. Patents 6,027,913 and 6,361,949.