missing translation for 'onlineSavingsMsg'
Learn More

Promega TNT™ SP6 High-Yield Protein Expression System

Catalog No. PRL3261
Encompass
Change view
Click to view available options
Quantity:
10 reactions
40 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRL3261 10 reactions
PRL3260 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRL3261 Supplier Promega Supplier No. L3261

Includes

TNT SP6 High-Yield Master Mix, Nuclease-free water

TNT SP6 High-Yield Wheat Germ Protein Expression System, based on an optimized wheat germ extract, is a single-tube, coupled transcription/translation system designed to express proteins in only two hours.

  • Coupled Transcription/Translation for Soluble, Full-Length Protein in Only Two Hours
  • Express soluble, full-length protein from plasmid or PCR-generated templates quickly
  • Generate 10- to 20-fold more protein (10-100μg/ml) than with other cell-free systems

Specifications

Kit Contents TnT™ SP6 high-yield master mix, Nuclease-free Water
Test Time 2 hr.
Yield 10 to 100 μg/mL
Type Wheat Germ Protein Expression System
Sufficient For 10 Reactions
Quantity 10 reactions
Content And Storage <-65°C

  • U.S. Patent 5,324,637; Australian Patent 660329 and other patents.
  • For Research Use. Any use of the product for diagnostics requiring clearance or approval by the FDA may require a license under Mayo Clinic U.S. Patents 6,027,913 and 6,361,949.