missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega TMB Stabilized Substrate for Horseradish Peroxidase
Description
- Premixed and ready to use
- Stable at room temperature for 12 months
- More sensitive and color-stable than 4-chloro-1-naphthol
Specifications
Specifications
| For Use With (Application) | Western blotting, dot blotting |
| Quantity | 200mL |
| Includes | Premixed and fully diluted in a proprietary buffer containing less than 0.5% organic solvent |
| pH | 5 |
| Storage Requirements | 15 to 30°C |
| Substrate Quantity | 200mL |