Learn More
Promega T7 RNA Polymerase DFS Item
DNA-dependent RNA polymerase for in vitro transcription.
Supplier:  Promega P2075
 View more versions of this product
Catalog No. PR-P2075
Only null left
Explore available promotions
Description
Description
  • DNA-Dependent RNA Polymerase Specific for T7 Promoter Sequences
  • >90% pure as determined by SDS polyacrylamide gel electrophoresis
  • Free of detectable levels of contaminating RNase and DNase activity (
  • Incorporates 32P, 33P, 3H and 35S nucleoside triphosphates
Specifications
Specifications
10 to 20 U/μL
5X Reaction Buffer: 100mM DTT and transcription optimized 5X Buffer: 200mM Tris HCl (pH 7.9 at 25°C), 30mM MgCl2, 10mM Spermidine, 50mM NaCl
7.7
20mM Potassium Phosphate (pH 7.7 at 25°C), 1mM EDTA, 10mM DTT, 100mM NaCl, 0.1% Triton™ X-100, 50% Glycerol
Recombinant E. coli strain
T7 RNA Polymerase
Liquid
Colorless
>90%
Generation of templates for in-vitro translation, generation of probes for Nucleic Acid hybridization, generation of RNA processing Substrates, generation of Antisense RNA
-30°C to -10°C
1000 U
Recombinant RNasin™ Ribonuclease
Product Suggestions
Product Suggestions