missing translation for 'onlineSavingsMsg'
Learn More

Promega T4 DNA Ligase Buffer Pack

Catalog No. PRC1263
Encompass
Change view
Click to view available options
Quantity:
3 x 500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRC1263 3 x 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRC1263 Supplier Promega Supplier No. C1263

For ligation of DNA ends.

  • Joins Two DNA Strands with Cohesive or Blunt Ends
  • Use with 5′, 3′ or blunt-ended dsDNA (e.g., inserts and vectors)
  • Qualified for blue/white cloning, providing a higher level of quality control for enzymes used in cloning applications
  • Can catalyze the joining of RNA to either a DNA or RNA strand in a duplex molecule

Specifications

Color Colorless
Components 3 Tubes of T4 DNA Ligase 10X Reaction Buffer: 300mM Tris HCl (pH 7.8 at 25degC), 100mM MgCl2, 100mM DTT and 10mM ATP
pH 7.8
For Use With (Application) For T4 DNA Kinase Enzyme
Content And Storage -30°C to -10°C
Quantity 3 x 500 μL
Product Type T4 DNA Ligase Buffer pack
Form Liquid