missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega T4 DNA Ligase
Description
- Joins Two DNA Strands with Cohesive or Blunt Ends
- Use with 5′, 3′ or blunt-ended dsDNA (e.g., inserts and vectors)
- Qualified for blue/white cloning, providing a higher level of quality control for enzymes used in cloning applications
- Can catalyze the joining of RNA to either a DNA or RNA strand in a duplex molecule
Specifications
Specifications
| Concentration | 1 to 3 U/μL |
| Color | Colorless |
| Components | 10X Reaction Buffer: 300mM Tris HCl (pH 7.8 at 25°C), 100mM MgCl2, 100mM DTT and 10mM ATP |
| Purity | >90% |
| pH | 7.4 |
| For Use With (Application) | Joining double-stranded DNA molecules with cohesive or blunt ends (5', 3' or blunt-ended DNA inserts) |
| Storage Buffer | 10mM Tris HCl (pH 7.4 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 50% Glycerol |
| Content And Storage | -30°C to -10°C |
| Source | Recombinant E. coli strain |
| Quantity | 100 U |
| Show More |