missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega SV 96 Total RNA Isolation System
The SV 96 Total RNA Isolation System provides a high-throughput technique to prepare intact RNA from tissue and cultured cells.
$739.17 - $2,245.77
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| For Use With (Application) | Preparation of intact, purified total RNA and RT-PCR |
Includes: Binding plate; DNA elution plate; DNase I (lyophilized); β-mercaptoethanol; SV DNase stop solution; MnCl2 (0.09M); Nuclease-free water; SV RNA wash solution; SV RNA lysis buffer; Yellow core buffer; Plate sealers
Requires: Use of the Vac-Man™ 96 Vacuum Manifold (Mfr. No. PR-A2291) or similar 96-well manifold for manual purification.
Description
- Purifies Total RNA From 96 Samples in Under an Hour
- Vacuum-based method. No centrifugation required
- Plate design prevents cross-contamination during sample processing
- Designed for both manual and automated RNA purification
Specifications
| 15°C to 30°C | |
| Preparation of intact, purified total RNA and RT-PCR |