missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega SP6 RNA Polymerase
Description
- DNA-Dependent RNA Polymerase Specific for SP6 Promoter Sequences
- Exhibits extremely high affinity and specificity for SP6 promoter sequences
- >90% pure as determined by SDS polyacrylamide gel electrophoresis
- Free of detectable levels of contaminating RNase and DNase activity (
Specifications
Specifications
| Concentration | 80 U/μL |
| Color | Colorless |
| Components | 100mM DTT 5X Reaction Buffer, transcription optimized 5X Buffer: 200mM Tris HCl (pH 7.9 at 25°C), 30mM MgCl2, 10mM Spermidine, 50mM NaCl |
| Purity | >90% |
| pH | 7.5 |
| For Use With (Application) | Generation of templates for in-vitro translation, generation of probes for Nucleic Acid hybridization, generation of RNA processing Substrates, generation of Antisense RNA |
| Storage Buffer | 20mM Potassium Phosphate (pH 7.7 at 25°C), 1mM EDTA, 10mM DTT, 100mM NaCl, 0.1% Triton™ X-100, 50% Glycerol |
| Content And Storage | -30°C to -10°C |
| Source | Recombinant E. coli strain |
| Quantity | 2500 U |
| Show More |