Learn More
Promega SoftLink™ Soft Release Avidin Resin
SoftLink Avidin Resin is used for the isolation and purification of biotinylated molecules.
Supplier: Promega V2012
Description
- Reversible Binding of Biotinylated Proteins Including Antibodies
- Specific capture of biotinylated proteins with reversible binding under mild elution conditions
- Binding ability retained under a wide range of pHs, and low or high ionic strength solutions
- Regenerates at least 10 times without loss of binding capacity
- For research use only. Not for diagnostic or therapeutic use.
- Exclusively licensed under U.S. Patents 5,395,856; and 6,072,039 and other patents.
Specifications
| 300 cm/hr. | |
| Methacrylate Polymeric Gel filtration matrix | |
| 5 mL | |
| Resin |
| 2 to 13 | |
| 2°C to 10°C | |
| Isolation and purification of Biotinylated Antibodies and other Proteins |