missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Sequencing Grade Modified Trypsin
Description
- Manufactured for Maximum Specificity and Stability
- Lysine residues modified by reductive methylation, yielding a highly active and stable trypsin
- Available lyophilized or frozen
- Referenced in thousands of papers
Specifications
Specifications
| Concentration | 0.1 mg/mL |
| Color | White |
| pH | 7.7 |
| For Use With (Application) | 'Hydrolyzing the Peptide at the Carboxyl side of Lysine and Arginine residues |
| Storage Buffer | 50mM Acetic Acid Resuspension Buffer |
| Content And Storage | -30°C to -10°C |
| Quantity | 100 μg |
| Product Type | Trypsin |
| Form | Lyophilized Powder |