missing translation for 'onlineSavingsMsg'
Learn More

Promega Sequencing Grade Chymotrypsin

Catalog No. PRV1062
Encompass
Change view
Click to view available options
Quantity:
100 μg
25 μg
Concentration:
0.5 to 1 μg/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PRV1062 100 μg 0.5 to 1 μg/μL
PRV1061 25 μg 0.5 to 1 μg/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1062 Supplier Promega Supplier No. V1062

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chymotrypsin is a highly-purified serine endopeptidase derived from bovine pancreas that preferentially hydrolyzes at the carboxyl side of aromatic amino acids: Tyr, Phe and Trp. Cleavage may also be observed, but at a lower rate, at Leu and Met.

  • Serine Protease that Cleaves at C-Terminus of Tyr, Phe and Trp
  • Optimal activity at pH 7.0-9.0.
  • Use alone or with other proteases for peptide mapping, peptide mass fingerprinting, MS/MS spectral matching
  • Use for digestion reactions in-solution or in-gel

Specifications

Concentration 0.5 to 1 μg/μL
Color Colorless
Purity >90%
pH 7 to 9
For Use With (Application) Cleaves Peptide bonds at the C-termini of Tyr, Phe, Trp and Leu. Met, Ala, Asp and Glu may be cleaved at a much lower rate
Content And Storage 2°C to 10°C
Source Bovine pancreas
Quantity 100 μg
Product Type Chymotrypsin
Molecular Weight (g/mol) 25
Form Lyophilized Powder
Show More Show Less