missing translation for 'onlineSavingsMsg'
Learn More

Promega ADP-Glo™ Kinase Assay + RSK2 Kinase Enzyme System

Catalog No. PRV9651
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9651 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV9651 Supplier Promega Supplier No. V9651

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant full-length human RSK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RSK2 is a member of the RSK (ribosomal S6 kinase) family that are growth factor-regulated serine-threonine kinases.

  • Easily Screen and Profile RSK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type Assay System
Includes Kinase, Substrate, Reaction Buffer, DTT
Content And Storage Components at different temps
For Use With (Application) Gene expression activation
Quantity 1/Ea.