missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega ROCK1 Kinase Enzyme System
Description
- Easily Screen and Profile ROCK1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Includes | 1 x 10μg ROCK1 Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay |
| Molecular Weight (g/mol) | 85 |
| Content And Storage | Components at different temps |
| For Use With (Application) | Diverse cellular functions, including smooth muscle contraction, actin cytoskeleton organization, cell adhesion and motility and gene expression |
| Quantity | 1/Ea. |
| Form | Fluid |