missing translation for 'onlineSavingsMsg'
Learn More

Promega ADP-Glo™ Kinase Assay + p70S6K Kinase Enzyme System

Catalog No. prv9611
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9611 1000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV9611 Supplier Promega Supplier No. V9611

Recombinant full-length human p70S6K was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. p70S6K is responsible for the phosphorylation of 40S ribosomal protein S6 and is ubiquitously expressed in human adult tissues.

  • Easily Screen and Profile p70S6K Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg p70S6K Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 76
Content And Storage Components at different temps
Quantity 1000 Assays
Form Fluid