Learn More
Promega Restriction Enzymes - Sac I Blue/White Cloning Qualified (J)
Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. SacI recognizes the same DNA sequence but cleaves at a different nucleotide than its neoschizomer, EcoICRI. (G AGCT/C C\TCGA G)
Supplier: Promega R6065
Description
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
Specifications
| 10 U/μL | |
| 37°C | |
| Provides a higher level of quality control for Enzymes used in cloning applications | |
| -30°C to -10°C | |
| GAGCT.C C.TCGAG | |
| Liquid |
| 1 x 1mL 10X Buffer J, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U SacI, 1 x 0.25mL Multi-Core™ 10X Buffer | |
| 7.4 | |
| 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
| 5000 U | |
| SacI |