missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Restriction Enzymes - Sac I Blue/White Cloning Qualified (J)
Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. SacI recognizes the same DNA sequence but cleaves at a different nucleotide than its neoschizomer, EcoICRI. (G AGCT/C C\TCGA G)
$82.79 - $302.61
Specifications
| Incubator Temperature | 37°C |
|---|---|
| pH | 7.4 |
| For Use With (Application) | Provides a higher level of quality control for Enzymes used in cloning applications |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
| Cut Site | GAGCT.C C.TCGAG |
Description
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
| 37°C | |
| Provides a higher level of quality control for Enzymes used in cloning applications | |
| GAGCT.C C.TCGAG |
| 7.4 | |
| 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
| Liquid |