Learn More
Promega Nuclei Lysis Solution for Wizard™ Genomic DNA Purification Kits
Component of Wizard, Wizard SV and Wizard SV 96 Genomic DNA Purification Systems, which are intended for isolating genomic DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.
Supplier: Promega A7963
Description
- Manual, Solution-Based DNA Extraction Method
- DNA from blood, animal and plant cells, bacteria or yeast
- 60-minute, scalable protocol
- Kit size based on number of isolations from whole blood, other sample types may vary
Identifiants chimiques
| Wizard™ genomic DNA purification kit | |
| Isolation of DNA from white Blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram positive and gram negative bacteria |
Spécifications
| 15°C to 30°C | |
| Isolation of DNA from white Blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram positive and gram negative bacteria | |
| 50 mL |
| Wizard™ genomic DNA purification kit | |
| DNA Rehydration Solution |