Learn More
Promega Reagents for ReliaPrep™ 96 gDNA Miniprep HT System DFS Item
Tris-EDTA and Tris-HCl buffer solutions that are quality tested and certified free of DNase and RNase activity.
$40.92 - $52.33
Specifications
For Use With (Equipment) ReliaPrep™ 96 genomic DNA Miniprep HT System
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRA2651 DFS Item
View Documents
Promega
A2651
25 mL
Each for $52.33
Only null left
PRA2641 DFS Item
View Documents
Promega
A2641
60 mL
Each for $40.92
Only null left
Description
Description
  • Mfr. No. A2641 is a component of Maxwell™ HT 96 gDNA Blood Isolation System (Mfr. No. A2670, A2671)
  • Mfr. No. A2651 is a buffer composed of 0.2mM Tris-HCl and 20mM EDTA (pH 7.5 at 25°C)
Specifications
Specifications
ReliaPrep™ 96 genomic DNA Miniprep HT System