missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega PureYield™ Plasmid Miniprep System
The PureYield Plasmid Miniprep System is designed to rapidly isolate highly pure plasmid DNA. Yields up to 15ug of plasmid DNA from 600ul to 3ml of bacterial culture.
$277.87 - $3,080.73
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| For Use With (Application) | Isolates transfection-quality plasmid DNA directly from culture in as little as 10 min. |
Includes: Plasmid Miniprep System: Cell lysis buffer, column wash solution, elution buffer, endotoxin removal wash, neutralization solution, PureYield collection tubes, PureYield minicolumns
Plasmid Miniprep Start-Up Kit (110V): Plasmid Miniprep System, Welch™ Vacuum Pump for Continental Europe Electrical, Vac-Man™ Laboratory Vacuum Manifold (20-sample capacity)
Description
- Up to 15μg of Transfection-Ready Plasmid from 3mL Cultures
- Rapid, 10-minute protocol
- Highly pure, concentrated plasmid DNA
- No tedious, gravity-drip columns or post-elution alcohol precipitation
Specifications
| 15°C to 30°C | |
| Isolates transfection-quality plasmid DNA directly from culture in as little as 10 min. |
U.S. Patent 6,194,562, Australian Patent 740145, Canadian Patent 2,329,067.